Assay Details
| Gene Symbol: mRpL13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078874.4 | NP_523598.2 | 2 - 3 | 242 | 65 | |
| GenBank mRNA | BT053702.1 | - | 2 - 3 | 184 | 65 | |
More Information
Set Membership: |
|

mRpL13
Genomic Map