Assay Details
| Gene Symbol: RpL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080080.3 | NP_524819.1 | 4 - 5 | 383 | 59 | |
| NM_166655.2 | NP_726412.1 | 3 - 4 | 375 | 59 | ||
| NM_166656.2 | NP_726413.1 | 4 - 5 | 383 | 59 | ||
| GenBank mRNA | AY071246.1 | - | 4 - 5 | 362 | 59 | |
| KX532072.1 | - | 3 - 4 | 291 | 59 | ||
More Information
Set Membership: |
|

RpL12
Genomic Map