Assay Details
| Gene Symbol: MED26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143679.4 | NP_651936.1 | 3 - 4 | 1125 | 83 | |
| NM_166791.2 | NP_726610.1 | 3 - 4 | 1046 | 83 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MED26| Gene Symbol: MED26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143679.4 | NP_651936.1 | 3 - 4 | 1125 | 83 | |
| NM_166791.2 | NP_726610.1 | 3 - 4 | 1046 | 83 | ||
Set Membership: |
|