Assay Details
| Gene Symbol: MED26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143679.4 | NP_651936.1 | 5 - 6 | 1829 | 137 | |
| NM_166791.2 | NP_726610.1 | 5 - 6 | 1750 | 137 | ||
More Information
Set Membership: |
|

MED26
Genomic Map