Assay Details
| Gene Symbol: Zyx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258501.3 | NP_001245430.1 | 7 - 8 | 1751 | 80 | |
| NM_143758.4 | NP_652015.1 | 7 - 8 | 1854 | 80 | ||
| NM_166815.3 | NP_726638.1 | 6 - 7 | 1581 | 80 | ||
| NM_166816.3 | NP_726639.1 | 7 - 8 | 1772 | 80 | ||
| NM_166819.3 | NP_726642.1 | 8 - 9 | 1973 | 80 | ||
| GenBank mRNA | AF219948.1 | - | 7 - 8 | 1807 | 80 | |
| AY069355.1 | - | 7 - 8 | 1744 | 80 | ||
| BT150084.1 | - | 6 - 7 | 1550 | 80 | ||
More Information
Set Membership: |
|

Zyx
Genomic Map