Assay Details
Gene Symbol: bgcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299858.1 | NP_001286787.1 | 3 - 4 | 3034 | 82 | |
NM_079108.3 | NP_523832.2 | 3 - 4 | 3049 | 82 | ||
GenBank mRNA | AF255662.1 | - | 3 - 4 | 2919 | 82 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
