Assay Details
Gene Symbol: bgcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299858.1 | NP_001286787.1 | 4 - 5 | 3717 | 108 | |
NM_079108.3 | NP_523832.2 | 4 - 5 | 3732 | 108 | ||
GenBank mRNA | AF255662.1 | - | 4 - 5 | 3602 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
