Assay Details
| Gene Symbol: mRpL20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079327.3 | NP_524051.1 | 1 - 2 | 190 | 72 | |
| GenBank mRNA | BT089020.1 | - | 1 - 2 | 156 | 72 | |
More Information
Set Membership: |
|

mRpL20
Genomic Map