Assay Details
| Gene Symbol: Galphaf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079394.3 | NP_524118.1 | 4 - 5 | 839 | 61 | |
| GenBank mRNA | BT064643.1 | - | 4 - 5 | 822 | 61 | |
| L09700.1 | - | 4 - 5 | 833 | 61 | ||
More Information
Set Membership: |
|

Galphaf
Genomic Map