Assay Details
| Gene Symbol: Zdhhc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103451.3 | NP_001096921.1 | 4 - 5 | 1217 | 57 | |
| NM_001103452.4 | NP_001096922.1 | 4 - 5 | 1103 | 57 | ||
| NM_001258663.3 | NP_001245592.1 | 4 - 5 | 1217 | 57 | ||
| NM_001272453.1 | NP_001259382.1 | 4 - 5 | 1103 | 57 | ||
| NM_001298122.1 | NP_001285051.1 | 4 - 5 | 1231 | 57 | ||
| NM_001298123.1 | NP_001285052.1 | 4 - 5 | 1202 | 57 | ||
| NM_167189.6 | NP_727339.3 | 4 - 5 | 1103 | 57 | ||
More Information
Set Membership: |
|

Zdhhc8
Genomic Map