Assay Details
| Gene Symbol: Pgm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079936.3 | NP_524675.1 | 2 - 3 | 369 | 59 | |
| GenBank mRNA | AF416981.1 | - | 2 - 3 | 355 | 59 | |
| AF416984.1 | - | 2 - 3 | 355 | 59 | ||
| BT010043.1 | - | 2 - 3 | 349 | 59 | ||
More Information
Set Membership: |
|

Pgm1
Genomic Map