Assay Details
| Gene Symbol: lmgB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103654.2 | NP_001097124.1 | 1 | 2673 | 150 | |
| NM_001273330.1 | NP_001260259.1 | 1 | 2673 | 150 | ||
| Gene Symbol: lmgA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103652.2 | NP_001097122.1 | 1 | 2673 | 150 | |
| NM_001273332.1 | NP_001260261.1 | 1 | 2673 | 150 | ||
| GenBank mRNA | AJ251510.1 | - | 1 | 2663 | 150 | |
More Information
Set Membership: |
|

lmgB
Genomic Map