Assay Details
Gene Symbol: veil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299604.1 | NP_001286533.1 | 4 - 5 | 632 | 71 | |
NM_137373.3 | NP_611217.1 | 3 - 4 | 433 | 71 | ||
GenBank mRNA | AY069716.1 | - | 3 - 4 | 456 | 71 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
