Assay Details
Gene Symbol: veil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299604.1 | NP_001286533.1 | 6 - 7 | 1180 | 81 | |
NM_137373.3 | NP_611217.1 | 5 - 6 | 981 | 81 | ||
GenBank mRNA | AY069716.1 | - | 5 - 6 | 1004 | 81 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
