Assay Details
| Gene Symbol: Sik3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103899.2 | NP_001097369.2 | 2 - 3 | 1062 | 73 | |
| NM_001103900.2 | NP_001097370.2 | 2 - 3 | 1062 | 73 | ||
| NM_001202040.1 | NP_001188969.1 | 2 - 3 | 1244 | 73 | ||
| NM_001299658.1 | NP_001286587.1 | 2 - 3 | 1062 | 73 | ||
| NM_137515.3 | NP_611359.1 | 2 - 3 | 852 | 73 | ||
| NM_137517.5 | NP_611361.4 | 2 - 3 | 412 | 73 | ||
| GenBank mRNA | BT023509.1 | - | 2 - 3 | 1015 | 73 | |
More Information
Set Membership: |
|

Sik3
Genomic Map