Assay Details
| Gene Symbol: Stoml2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274232.1 | NP_001261161.1 | 6 - 7 | 897 | 60 | |
| NM_138009.3 | NP_611853.2 | 6 - 7 | 1050 | 60 | ||
| GenBank mRNA | BT024262.1 | - | 6 - 7 | 846 | 60 | |
More Information
Set Membership: |
|

Stoml2
Genomic Map