Assay Details
Gene Symbol: wac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169835.1 | NP_001163306.1 | 1 - 2 | 361 | 67 | |
NM_001274265.1 | NP_001261194.1 | 2 - 3 | 536 | 67 | ||
NM_138176.3 | NP_612020.2 | 2 - 3 | 362 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
