Assay Details
| Gene Symbol: feo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132430.3 | NP_572658.2 | 1 | 2335 | 122 | |
| GenBank mRNA | BT032843.1 | - | 1 | 2323 | 122 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

feo| Gene Symbol: feo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132430.3 | NP_572658.2 | 1 | 2335 | 122 | |
| GenBank mRNA | BT032843.1 | - | 1 | 2323 | 122 | |
Set Membership: |
|