Assay Details
Gene Symbol: Paics | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272564.1 | NP_001259493.1 | 4 - 5 | 726 | 70 | |
NM_132598.4 | NP_572826.1 | 4 - 5 | 729 | 70 | ||
GenBank mRNA | AF102579.1 | - | 4 - 5 | 711 | 70 | |
AY095520.1 | - | 4 - 5 | 694 | 70 | ||
BT126073.1 | - | 4 - 5 | 724 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
