Assay Details
| Gene Symbol: CG2691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298289.1 | NP_001285218.1 | 1 - 2 | 248 | 69 | |
| NM_132678.3 | NP_572906.2 | 1 - 2 | 248 | 69 | ||
More Information
Set Membership: |
|

CG2691
Genomic Map