Assay Details
| Gene Symbol: dap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057600.6 | NP_476948.1 | 1 - 2 | 786 | 62 | |
| GenBank mRNA | BT125118.1 | - | 1 - 2 | 462 | 62 | |
| U68477.1 | - | 1 - 2 | 798 | 62 | ||
More Information
Set Membership: |
|

dap
Genomic Map