Assay Details
Gene Symbol: SdhB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057753.5 | NP_477101.1 | 1 - 2 | 194 | 83 | |
GenBank mRNA | AY118923.1 | - | 1 - 2 | 191 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
