Assay Details
Gene Symbol: dpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299231.1 | NP_001286160.1 | 3 - 4 | 1680 | 61 | |
NM_057837.5 | NP_477185.1 | 3 - 4 | 1412 | 61 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
