Assay Details
| Gene Symbol: mRpL33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080242.4 | NP_524981.2 | 2 - 3 | 242 | 68 | |
| GenBank mRNA | AY118791.1 | - | 2 - 3 | 212 | 68 | |
More Information
Set Membership: |
|

mRpL33
Genomic Map