Assay Details
Gene Symbol: Rbf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080297.3 | NP_525036.2 | 2 - 3 | 1027 | 60 | |
GenBank mRNA | AY089532.1 | - | 2 - 3 | 1005 | 60 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
