Assay Details
Gene Symbol: Rok | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298406.1 | NP_001285335.1 | 2 - 3 | 1130 | 69 | |
NM_001298407.1 | NP_001285336.1 | 2 - 3 | 1130 | 69 | ||
NM_080535.3 | NP_536796.2 | 2 - 3 | 1130 | 69 | ||
GenBank mRNA | BT044181.1 | - | 2 - 3 | 1135 | 69 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
