Assay Details
Gene Symbol: Rok | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298406.1 | NP_001285335.1 | 5 - 6 | 4159 | 64 | |
NM_001298407.1 | NP_001285336.1 | 5 - 6 | 4159 | 64 | ||
NM_001298408.1 | NP_001285337.1 | 4 - 5 | 3160 | 64 | ||
NM_080535.3 | NP_536796.2 | 5 - 6 | 4159 | 64 | ||
GenBank mRNA | BT044181.1 | - | 5 - 6 | 4164 | 64 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
