Assay Details
Gene Symbol: Rok | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298407.1 | NP_001285336.1 | 6 - 7 | 4437 | 71 | |
NM_001298408.1 | NP_001285337.1 | 5 - 6 | 3438 | 71 | ||
NM_080535.3 | NP_536796.2 | 6 - 7 | 4437 | 71 | ||
GenBank mRNA | BT044181.1 | - | 6 - 7 | 4442 | 71 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
