Assay Details
| Gene Symbol: STUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058093.4 | NP_477441.1 | 2 - 3 | 619 | 64 | |
| GenBank mRNA | AF129084.1 | - | 2 - 3 | 568 | 64 | |
| AY069760.1 | - | 2 - 3 | 612 | 64 | ||
More Information
Set Membership: |
|

STUB1
Genomic Map