Assay Details
Gene Symbol: Hel25E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273171.1 | NP_001260100.1 | 3 - 4 | 614 | 84 | |
NM_164646.3 | NP_723089.1 | 3 - 4 | 614 | 84 | ||
NM_164647.1 | NP_723090.1 | 2 - 3 | 504 | 84 | ||
NM_164648.2 | NP_723091.1 | 3 - 4 | 607 | 84 | ||
GenBank mRNA | AY118921.1 | - | 3 - 4 | 569 | 84 | |
X79802.1 | - | 585 | 84 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
