Assay Details
| Gene Symbol: Doa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014678.3 | NP_001014678.1 | 4 - 5 | 2229 | 71 | |
| NM_001014679.2 | NP_001014679.1 | 2 - 3 | 1592 | 71 | ||
| NM_001014680.4 | NP_001014680.1 | 5 - 6 | 945 | 71 | ||
| NM_001014681.4 | NP_001014681.2 | 4 - 5 | 5210 | 71 | ||
| NM_001014682.3 | NP_001014682.1 | 5 - 6 | 821 | 71 | ||
| NM_001043298.2 | NP_001036763.2 | 3 - 4 | 3655 | 71 | ||
| NM_001043299.3 | NP_001036764.2 | 6 - 7 | 1028 | 71 | ||
| NM_001043300.3 | NP_001036765.1 | 5 - 6 | 945 | 71 | ||
| NM_001144648.3 | NP_001138120.1 | 6 - 7 | 888 | 71 | ||
| NM_001144649.2 | NP_001138121.1 | 6 - 7 | 888 | 71 | ||
| NM_001144650.1 | NP_001138122.1 | 5 - 6 | 821 | 71 | ||
| NM_001260424.1 | NP_001247353.1 | 3 - 4 | 1866 | 71 | ||
| NM_001260425.1 | NP_001247354.1 | 4 - 5 | 1857 | 71 | ||
| NM_001382085.1 | NP_001368951.1 | 6 - 7 | 1085 | 71 | ||
| NM_176581.4 | NP_788758.3 | 4 - 5 | 2619 | 71 | ||
| GenBank mRNA | AY061474.1 | - | 5 - 6 | 939 | 71 | |
| AY069573.1 | - | 2 - 3 | 1592 | 71 | ||
| BT133441.1 | - | 4 - 5 | 2227 | 71 | ||
| EF489455.1 | - | 4 - 5 | 2227 | 71 | ||
More Information
Set Membership: |
|

Doa
Genomic Map