Assay Details
Gene Symbol: osa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170168.2 | NP_001163639.1 | 12 - 13 | 4345 | 59 | |
NM_079668.3 | NP_524392.2 | 12 - 13 | 4636 | 59 | ||
NM_169775.2 | NP_732263.1 | 12 - 13 | 4597 | 59 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
