Assay Details
Gene Symbol: osa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170168.2 | NP_001163639.1 | 6 - 7 | 2252 | 63 | |
NM_001275751.1 | NP_001262680.1 | 6 - 7 | 2252 | 63 | ||
NM_001275752.1 | NP_001262681.1 | 6 - 7 | 2543 | 63 | ||
NM_079668.3 | NP_524392.2 | 6 - 7 | 2543 | 63 | ||
NM_169775.2 | NP_732263.1 | 6 - 7 | 2543 | 63 | ||
NM_206506.2 | NP_996228.1 | 6 - 7 | 2543 | 63 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
