Assay Details
Gene Symbol: osa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170168.2 | NP_001163639.1 | 9 - 10 | 3424 | 66 | |
NM_001275751.1 | NP_001262680.1 | 9 - 10 | 3421 | 66 | ||
NM_079668.3 | NP_524392.2 | 9 - 10 | 3715 | 66 | ||
NM_169775.2 | NP_732263.1 | 9 - 10 | 3715 | 66 | ||
NM_206506.2 | NP_996228.1 | 9 - 10 | 3715 | 66 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
