Assay Details
| Gene Symbol: gpp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275387.1 | NP_001262316.1 | 4 - 5 | 2892 | 60 | |
| NM_141398.4 | NP_649655.1 | 5 - 6 | 2504 | 60 | ||
| NM_169142.2 | NP_731083.1 | 4 - 5 | 2890 | 60 | ||
| NM_169143.2 | NP_731084.1 | 5 - 6 | 2504 | 60 | ||
| NM_169144.2 | NP_731085.2 | 5 - 6 | 2504 | 60 | ||
More Information
Set Membership: |
|

gpp
Genomic Map