Assay Details
Gene Symbol: RhoGAP93B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142682.2 | NP_650939.2 | 10 - 11 | 3929 | 98 | |
GenBank mRNA | BT053685.1 | - | 10 - 11 | 3907 | 98 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
