Assay Details
| Gene Symbol: RhoGAP93B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142682.2 | NP_650939.2 | 11 - 12 | 4048 | 67 | |
| GenBank mRNA | BT053685.1 | - | 11 - 12 | 4026 | 67 | |
More Information
Set Membership: |
|

RhoGAP93B
Genomic Map