Assay Details
| Gene Symbol: Cad96Ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143092.3 | NP_651349.1 | 3 - 4 | 607 | 90 | |
| GenBank mRNA | AY070958.1 | - | 3 - 4 | 605 | 90 | |
| BT001657.1 | - | 3 - 4 | 605 | 90 | ||
More Information
Set Membership: |
|

Cad96Ca
Genomic Map