Assay Details
| Gene Symbol: LpR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104456.2 | NP_001097926.1 | 9 - 10 | 3137 | 58 | |
| NM_001104457.2 | NP_001097927.1 | 5 - 6 | 1190 | 58 | ||
| NM_001104458.2 | NP_001097928.1 | 10 - 11 | 3242 | 58 | ||
| NM_001104459.2 | NP_001097929.1 | 9 - 10 | 2701 | 58 | ||
| NM_001104460.2 | NP_001097930.1 | 10 - 11 | 3239 | 58 | ||
| NM_001276041.1 | NP_001262970.1 | 9 - 10 | 2690 | 58 | ||
| NM_001276042.1 | NP_001262971.1 | 8 - 9 | 2585 | 58 | ||
| NM_170239.4 | NP_733118.3 | 9 - 10 | 3134 | 58 | ||
| NM_176565.2 | NP_788742.1 | 8 - 9 | 2529 | 58 | ||
| GenBank mRNA | BT010074.1 | - | 10 - 11 | 3220 | 58 | |
| BT010094.1 | - | 9 - 10 | 2612 | 58 | ||
| BT011437.1 | - | 5 - 6 | 1191 | 58 | ||
More Information
Set Membership: |
|

LpR2
Genomic Map