Assay Details
| Gene Symbol: AGO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043162.3 | NP_001036627.2 | 2 - 3 | 500 | 123 | |
| NM_001043163.3 | NP_001036628.2 | 2 - 3 | 473 | 123 | ||
| NM_001043164.3 | NP_001036629.2 | 2 - 3 | 674 | 123 | ||
| NM_001170027.2 | NP_001163498.1 | 2 - 3 | 500 | 123 | ||
| GenBank mRNA | EF434174.1 | - | 1 - 2 | 373 | 123 | |
More Information
Set Membership: |
|

AGO3
Genomic Map