Assay Details
| Gene Symbol: rl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015122.3 | NP_001015122.1 | 2 - 3 | 288 | 142 | |
| NM_001015123.3 | NP_001015123.1 | 1 - 2 | 469 | 142 | ||
| NM_001110878.4 | NP_001104348.2 | 2 - 3 | 264 | 142 | ||
| NM_001110879.2 | NP_001104349.1 | 2 - 3 | 247 | 142 | ||
| NM_001110880.2 | NP_001104350.1 | 1 - 2 | 469 | 142 | ||
| NM_001300706.2 | NP_001287635.1 | 2 - 3 | 264 | 142 | ||
| GenBank mRNA | AY070996.1 | - | 2 - 3 | 288 | 142 | |
| BT120325.1 | - | 2 - 3 | 200 | 142 | ||
| M95124.1 | - | 1 - 2 | 414 | 142 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001110878.4 transcripts.
Set Membership: |
|

rl
Genomic Map