Assay Details
| Gene Symbol: rl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015121.3 | NP_001015121.3 | 4 - 5 | 479 | 144 | |
| NM_001015122.3 | NP_001015122.1 | 3 - 4 | 410 | 144 | ||
| NM_001015123.3 | NP_001015123.1 | 2 - 3 | 591 | 144 | ||
| NM_001110879.2 | NP_001104349.1 | 3 - 4 | 369 | 144 | ||
| NM_001110880.2 | NP_001104350.1 | 2 - 3 | 591 | 144 | ||
| NM_001300706.2 | NP_001287635.1 | 3 - 4 | 386 | 144 | ||
| GenBank mRNA | AY070996.1 | - | 3 - 4 | 410 | 144 | |
| BT120325.1 | - | 3 - 4 | 322 | 144 | ||
| M95124.1 | - | 2 - 3 | 536 | 144 | ||
More Information
Set Membership: |
|

rl
Genomic Map