Assay Details
| Gene Symbol: rl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015121.3 | NP_001015121.3 | 7 - 8 | 1017 | 86 | |
| NM_001015122.3 | NP_001015122.1 | 6 - 7 | 948 | 86 | ||
| NM_001015123.3 | NP_001015123.1 | 5 - 6 | 1129 | 86 | ||
| NM_001110878.4 | NP_001104348.2 | 7 - 8 | 986 | 86 | ||
| NM_001110879.2 | NP_001104349.1 | 6 - 7 | 907 | 86 | ||
| NM_001300706.2 | NP_001287635.1 | 6 - 7 | 924 | 86 | ||
| GenBank mRNA | AY070996.1 | - | 6 - 7 | 948 | 86 | |
| M95124.1 | - | 5 - 6 | 1074 | 86 | ||
More Information
Set Membership: |
|

rl
Genomic Map