Assay Details
| Gene Symbol: abhd17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002744.1 | NP_001002744.1 | 2 - 3 | 801 | 65 | |
| GenBank mRNA | BC076440.1 | - | 2 - 3 | 801 | 65 | |
More Information
Set Membership: |
|

abhd17b
Genomic Map