Assay Details
| Gene Symbol: siae | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044331.1 | NP_001037796.1 | 5 - 6 | 754 | 68 | |
| GenBank mRNA | BC049448.1 | - | 5 - 6 | 754 | 68 | |
| BC134862.1 | - | 5 - 6 | 726 | 68 | ||
More Information
Set Membership: |
|

siae
Genomic Map