Assay Details
| Gene Symbol: cfap126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045418.1 | NP_001038883.1 | 2 - 3 | 133 | 104 | |
| GenBank mRNA | BC122100.1 | - | 2 - 3 | 133 | 104 | |
More Information
Set Membership: |
|

cfap126
Genomic Map