Assay Details
| Gene Symbol: hps3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193542.1 | NP_001180471.1 | 1 - 2 | 344 | 68 | |
| XM_005161859.2 | XP_005161916.1 | 1 - 2 | 345 | 68 | ||
| GenBank mRNA | BC124612.1 | - | 1 - 2 | 351 | 68 | |
| BC165396.1 | - | 1 - 2 | 253 | 68 | ||
More Information
Set Membership: |
|

hps3
Genomic Map