Assay Details
| Gene Symbol: gls2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083825.1 | NP_001077294.1 | 3 - 4 | 441 | 63 | |
| XM_005173960.4 | XP_005174017.1 | 3 - 4 | 968 | 63 | ||
| GenBank mRNA | BC134927.1 | - | 3 - 4 | 441 | 63 | |
More Information
Set Membership: |
|

gls2b
Genomic Map