Assay Details
| Gene Symbol: yipf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199907.1 | NP_956201.1 | 7 - 8 | 900 | 62 | |
| GenBank mRNA | BC044136.1 | - | 7 - 8 | 900 | 62 | |
| BC164802.1 | - | 7 - 8 | 808 | 62 | ||
More Information
Set Membership: |
|

yipf3
Genomic Map