Assay Details
| Gene Symbol: irf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200598.2 | NP_956892.1 | 3 - 4 | 643 | 84 | |
| GenBank mRNA | AY725802.2 | - | 3 - 4 | 643 | 84 | |
| BC056772.1 | - | 3 - 4 | 597 | 84 | ||
More Information
Set Membership: |
|

irf6
Genomic Map